siRNA Details
Gene Symbol: GRXCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080516.1 | NP_001073985.1 | 2 | 370 | ||
XM_017009708.1 | XP_016865197.1 | 2 | 210 | |||
GenBank mRNA | DN831963.1 | - | 2 | 308 |
