siRNA Details
Gene Symbol: ZNF862 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099220.1 | NP_001092690.1 | 7 | 2416 | ||
XM_006716085.3 | XP_006716148.1 | 8 | 2437 | |||
XM_011516463.1 | XP_011514765.1 | 7 | 3297 | |||
XM_011516464.1 | XP_011514766.1 | 7 | 3686 | |||
XM_011516465.2 | XP_011514767.1 | 4 | 1351 | |||
XM_011516466.2 | XP_011514768.1 | 3 | 1194 | |||
XM_017012519.1 | XP_016868008.1 | 7 | 2426 | |||
GenBank mRNA | AB011115.1 | - | 5 | 2051 | ||
DA724031.1 | - | 1 | 546 |
