siRNA Details
Gene Symbol: SDHAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042631.2 | NP_001036096.1 | 1 | 529 | ||
GenBank mRNA | BC007205.1 | - | 1 | 518 | ||
BC137516.1 | - | 1 | 472 | 9021516 | ||
BC137517.1 | - | 1 | 472 | 9021517 |
