siRNA Details
Gene Symbol: PIRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101387.1 | NP_001094857.1 | 2 | 339 | ||
GenBank mRNA | AK021549.1 | - | 2 | 320 | ||
AK299808.1 | - | 2 | 325 | |||
DC332889.1 | - | 2 | 339 |
