siRNA Details
Gene Symbol: DEFB131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040448.2 | NP_001035538.2 | 2 | 70 | ||
GenBank mRNA | AF529414.1 | - | 2 | 70 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DEFB131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040448.2 | NP_001035538.2 | 2 | 70 | ||
GenBank mRNA | AF529414.1 | - | 2 | 70 |