siRNA Details
Gene Symbol: LOC644554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040013.1 | 1, 2 | 300 | |||
GenBank mRNA | AK092337.1 | - | 1, 2 | 196 | ||
AK309685.1 | - | 1, 2 | 300 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC644554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040013.1 | 1, 2 | 300 | |||
GenBank mRNA | AK092337.1 | - | 1, 2 | 196 | ||
AK309685.1 | - | 1, 2 | 300 |