siRNA Details
Gene Symbol: LINC01091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027105.2 | 2 | 148 | |||
GenBank mRNA | AL833449.1 | - | 3 | 213 | ||
CK821624.1 | - | 2 | 152 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027105.2 | 2 | 148 | |||
GenBank mRNA | AL833449.1 | - | 3 | 213 | ||
CK821624.1 | - | 2 | 152 |