siRNA Details
Gene Symbol: ACTG1P20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033926.1 | 1 | 2280 | |||
GenBank mRNA | AK056682.1 | - | 1 | 2280 | ||
BX648289.1 | - | 1 | 3758 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ACTG1P20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033926.1 | 1 | 2280 | |||
GenBank mRNA | AK056682.1 | - | 1 | 2280 | ||
BX648289.1 | - | 1 | 3758 |