siRNA Details
Gene Symbol: CFAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304548.1 | NP_001291477.1 | 62 | 9150 | ||
GenBank mRNA | BG190566.1 | - | 2 | 257 | ||
BG197183.1 | - | 2 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
