siRNA Details
Gene Symbol: LINC00693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038840.1 | 5 | 825 | |||
GenBank mRNA | AA434563.1 | - | 3 | 361 | ||
BX333929.2 | - | 3 | 505 | |||
R20280.1 | - | 2 | 287 | |||
R20979.1 | - | 2 | 300 |
