siRNA Details
Gene Symbol: LINC01572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126330.1 | 5 | 308 | |||
GenBank mRNA | AL525401.3 | - | 5 | 248 | ||
AL706640.1 | - | 4 | 274 | |||
DA877605.1 | - | 6 | 394 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126330.1 | 5 | 308 | |||
GenBank mRNA | AL525401.3 | - | 5 | 248 | ||
AL706640.1 | - | 4 | 274 | |||
DA877605.1 | - | 6 | 394 |