siRNA Details
Gene Symbol: WFDC21P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030732.1 | 3 | 481 | |||
GenBank mRNA | AF086204.1 | - | 3 | 307 | ||
KJ020271.1 | - | 3 | 278 | |||
KJ020272.1 | - | 2 | 258 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WFDC21P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030732.1 | 3 | 481 | |||
GenBank mRNA | AF086204.1 | - | 3 | 307 | ||
KJ020271.1 | - | 3 | 278 | |||
KJ020272.1 | - | 2 | 258 |