siRNA Details
Gene Symbol: ZNF541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277075.1 | NP_001264004.1 | 3 | 1268 | ||
XM_005259311.4 | XP_005259368.1 | 4 | 2208 | |||
XM_011527367.2 | XP_011525669.1 | 4 | 2223 | |||
XM_011527368.2 | XP_011525670.1 | 5 | 1561 | |||
XM_011527369.2 | XP_011525671.1 | 5 | 1565 | |||
XM_011527370.2 | XP_011525672.1 | 4 | 1422 | |||
XM_011527371.2 | XP_011525673.1 | 4 | 1409 | |||
XM_011527372.2 | XP_011525674.1 | 4 | 2223 | |||
XM_011527373.2 | XP_011525675.1 | 4 | 2223 | |||
XM_011527374.2 | XP_011525676.1 | 4 | 2058 | |||
XM_011527375.1 | XP_011525677.1 | 4 | 2223 | |||
XM_017027351.1 | XP_016882840.1 | 4 | 2223 | |||
GenBank mRNA | AL833862.1 | - | 1 | 40 |
