siRNA Details
Gene Symbol: GFY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195256.1 | NP_001182185.1 | 2, 3 | 1349 | ||
XM_011526299.2 | XP_011524601.1 | 3, 4 | 1461 | |||
GenBank mRNA | DN930573.1 | - | 2, 3 | 204 |
