siRNA Details
Gene Symbol: C19orf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277378.1 | NP_001264307.1 | 1 | 418 | ||
XM_011528184.2 | XP_011526486.1 | 1 | 776 | |||
GenBank mRNA | HY028687.1 | - | 1 | 419 |
