siRNA Details
Gene Symbol: FABP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105281.1 | NP_001098751.1 | 3 | 312 | ||
XM_005251285.4 | XP_005251342.1 | 4 | 981 | |||
XM_006716465.3 | XP_006716528.1 | 4 | 964 | |||
XM_011517577.2 | XP_011515879.1 | 5 | 1108 | |||
GenBank mRNA | BC146973.1 | - | 3 | 375 | 9056901 | |
BC146981.1 | - | 3 | 375 | 9056909 | ||
EU733650.1 | - | 3 | 323 |
