siRNA Details
Gene Symbol: C4orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080531.1 | NP_001074000.1 | 1 | 113 | ||
XM_006714289.3 | XP_006714352.1 | 1 | 214 | |||
XM_011532196.2 | XP_011530498.1 | 1 | 214 | |||
XM_011532197.2 | XP_011530499.1 | 1 | 214 | |||
XM_017008547.1 | XP_016864036.1 | 1 | 214 | |||
XM_017008548.1 | XP_016864037.1 | 1 | 214 | |||
XM_017008549.1 | XP_016864038.1 | 1 | 214 | |||
XM_017008550.1 | XP_016864039.1 | 1 | 214 | |||
XM_017008551.1 | XP_016864040.1 | 1 | 214 | |||
XM_017008552.1 | XP_016864041.1 | 1 | 214 | |||
XM_017008553.1 | XP_016864042.1 | 1 | 214 | |||
XM_017008554.1 | XP_016864043.1 | 1 | 214 | |||
XM_017008555.1 | XP_016864044.1 | 1 | 214 | |||
XM_017008556.1 | XP_016864045.1 | 1 | 214 |
