siRNA Details
Gene Symbol: MROH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163629.1 | NP_001157101.1 | 21 | 2465 | ||
XM_011510005.2 | XP_011508307.1 | 20 | 2315 | |||
XM_011510006.2 | XP_011508308.1 | 20 | 2291 | |||
XM_011510007.2 | XP_011508309.1 | 11 | 1558 |
