siRNA Details
Gene Symbol: LOC646938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036495.1 | 1, 2 | 206 | |||
GenBank mRNA | BC127919.1 | - | 1, 2 | 136 | ||
BC141940.1 | - | 1, 2 | 136 | 40132506 | ||
BC150505.1 | - | 1, 2 | 136 | 40132514 |
