siRNA Details
Gene Symbol: KRTAP21-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB096960.1 | - | 1 | 355 |
Gene Symbol: LOC101927016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535339.1 | XP_011533641.1 | 1 | 1143 |
