siRNA Details
Gene Symbol: PRAMEF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099851.3 | NP_001093321.1 | 2 | 839 | ||
GenBank mRNA | BC144662.1 | - | 2 | 839 | 9053193 | |
BC146858.1 | - | 2 | 839 | 9056786 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
