siRNA Details
Gene Symbol: TMEM253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146683.1 | NP_001140155.1 | 8 | 1433 | ||
XM_006720233.3 | XP_006720296.1 | 7 | 2008 | |||
XM_011537078.2 | XP_011535380.1 | 8 | 1310 | |||
XM_011537079.1 | XP_011535381.1 | 7 | 1152 | |||
XM_011537081.2 | XP_011535383.1 | 6 | 1899 | |||
XM_011537082.2 | XP_011535384.1 | 7 | 1317 | |||
XM_011537083.2 | XP_011535385.1 | 6 | 1900 | |||
XM_011537084.2 | XP_011535386.1 | 6 | 1872 | |||
XM_011537085.2 | XP_011535387.1 | 5 | 1792 |
