siRNA Details
Gene Symbol: GGCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000821.6 | NP_000812.2 | 1, 2 | 152 | ||
NM_001311312.1 | NP_001298241.1 | 1, 2 | 127 | |||
XM_005264259.4 | XP_005264316.1 | 1, 2 | 111 | |||
XM_011532764.2 | XP_011531066.1 | 1, 2 | 111 | |||
GenBank mRNA | AK222827.1 | - | 1, 2 | 112 | ||
AK225663.1 | - | 1, 2 | 111 | |||
AK310483.1 | - | 1, 2 | 91 | |||
AK313126.1 | - | 1, 2 | 120 | |||
BC013979.2 | - | 1, 2 | 74 | 4129593 | ||
BM704752.1 | - | 1, 2 | 127 | |||
DC296316.1 | - | 1, 2 | 153 | |||
EU847509.1 | - | 1, 2 | 49 | |||
L17128.1 | - | 1, 2 | 201 | |||
M81592.1 | - | 1, 2 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
