siRNA Details
Gene Symbol: ANAPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022662.3 | NP_073153.1 | 35, 36 | 4629 | ||
XM_011511633.2 | XP_011509935.1 | 35, 36 | 4673 | |||
XM_011511634.2 | XP_011509936.1 | 35, 36 | 4788 | |||
XM_017004710.1 | XP_016860199.1 | 35, 36 | 4673 | |||
XM_017004711.1 | XP_016860200.1 | 35, 36 | 4661 | |||
XM_017004712.1 | XP_016860201.1 | 35, 36 | 4776 | |||
XM_017004713.1 | XP_016860202.1 | 34, 35 | 4736 | |||
XM_017004714.1 | XP_016860203.1 | 34, 35 | 4672 | |||
GenBank mRNA | AJ278357.1 | - | 35, 36 | 4644 | ||
AK023807.1 | - | 22, 23 | 2855 | |||
AL832013.1 | - | 24, 25 | 2985 | |||
BC104902.1 | - | 35, 36 | 4431 | 8143905 | ||
BC104904.1 | - | 35, 36 | 4431 | 8143907 |
Gene Symbol: LOC102724642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011533233.2 | XP_011531535.2 | 18, 19 | 2393 |
