siRNA Details
Gene Symbol: LINC00871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102699.1 | 3 | 372 | |||
NR_102700.1 | 2 | 276 | ||||
NR_102701.1 | 6 | 472 | ||||
NR_102702.1 | 4 | 355 | ||||
GenBank mRNA | BG186771.1 | - | 3 | 373 | ||
BG188315.1 | - | 2 | 277 | |||
BM725328.1 | - | 4 | 329 | |||
BU194844.1 | - | 6 | 472 |
