siRNA Details
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270974.2 | NP_001257903.1 | 50 | 8630 | ||
XM_006721206.2 | XP_006721269.1 | 50 | 8524 | |||
XM_011523146.2 | XP_011521448.1 | 51 | 8659 | |||
XM_011523147.1 | XP_011521449.1 | 51 | 8626 | |||
XM_011523148.1 | XP_011521450.1 | 51 | 8699 | |||
XM_011523151.1 | XP_011521453.1 | 50 | 8556 | |||
XM_011523152.1 | XP_011521454.1 | 13 | 2383 | |||
XM_011523155.2 | XP_011521457.1 | 6 | 1384 | |||
XM_017023346.1 | XP_016878835.1 | 50 | 8802 | |||
XM_017023347.1 | XP_016878836.1 | 38 | 7041 | |||
XM_017023348.1 | XP_016878837.1 | 38 | 6781 |
Gene Symbol: HYDIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103556.1 | 41 | 7152 | |||
GenBank mRNA | AB058767.1 | - | 15 | 2884 |
