siRNA Details
Gene Symbol: FAM226A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026595.2 | 1 | 658 | |||
GenBank mRNA | BC025725.1 | - | 1 | 123 | ||
BC041962.1 | - | 1 | 586 |
Gene Symbol: FAM226B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026594.1 | 1 | 658 |
