siRNA Details
Gene Symbol: GLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000404.3 | NP_000395.2 | 16 | 2087 | ||
NM_001079811.2 | NP_001073279.1 | 16 | 1981 | |||
NM_001135602.2 | NP_001129074.1 | 13 | 1694 | |||
NM_001317040.1 | NP_001303969.1 | 17 | 2231 | |||
GenBank mRNA | AK130133.1 | - | 16 | 2015 | ||
AK222763.1 | - | 16 | 1978 | |||
AK223091.1 | - | 16 | 2003 | |||
AK298985.1 | - | 13 | 1606 | |||
AK300021.1 | - | 16 | 1914 | |||
AK300753.1 | - | 17 | 2147 | |||
AK312988.1 | - | 16 | 1974 | |||
BC007493.2 | - | 16 | 1989 | 2988086 | ||
BT007147.1 | - | 16 | 1942 | |||
M22590.1 | - | 16 | 1976 | |||
M27507.1 | - | 16 | 1992 | |||
M27508.1 | - | 13 | 1599 | |||
M34423.1 | - | 16 | 2002 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
