siRNA Details
Gene Symbol: GLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000404.3 | NP_000395.2 | 14 | 1590 | ||
NM_001079811.2 | NP_001073279.1 | 14 | 1484 | |||
NM_001135602.2 | NP_001129074.1 | 11 | 1197 | |||
NM_001317040.1 | NP_001303969.1 | 15 | 1734 | |||
GenBank mRNA | AK130133.1 | - | 14 | 1518 | ||
AK222763.1 | - | 14 | 1481 | |||
AK223091.1 | - | 14 | 1506 | |||
AK298985.1 | - | 11 | 1109 | |||
AK300021.1 | - | 14 | 1417 | |||
AK300753.1 | - | 15 | 1650 | |||
AK312988.1 | - | 14 | 1477 | |||
BC007493.2 | - | 14 | 1492 | 2988086 | ||
BT007147.1 | - | 14 | 1445 | |||
M22590.1 | - | 14 | 1479 | |||
M27507.1 | - | 14 | 1495 | |||
M27508.1 | - | 11 | 1102 | |||
M34423.1 | - | 14 | 1505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
