siRNA Details
Gene Symbol: LINC00316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103811.1 | 1, 2 | 173 | |||
GenBank mRNA | BG221750.1 | - | 1, 2 | 177 | ||
BG221754.1 | - | 1, 2 | 177 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103811.1 | 1, 2 | 173 | |||
GenBank mRNA | BG221750.1 | - | 1, 2 | 177 | ||
BG221754.1 | - | 1, 2 | 177 |