siRNA Details
Gene Symbol: LOC101060391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136151.1 | 2 | 258 | |||
GenBank mRNA | CD519625.1 | - | 1 | 56 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101060391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136151.1 | 2 | 258 | |||
GenBank mRNA | CD519625.1 | - | 1 | 56 |