siRNA Details
Gene Symbol: KRTAP2-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165252.1 | NP_001158724.1 | 1 | 557 | ||
GenBank mRNA | BC063625.1 | - | 1 | 557 | 3907481 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRTAP2-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165252.1 | NP_001158724.1 | 1 | 557 | ||
GenBank mRNA | BC063625.1 | - | 1 | 557 | 3907481 |