siRNA Details
Gene Symbol: LOC728485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294306.1 | NP_001281235.1 | 2 | 297 | ||
GenBank mRNA | AK092138.1 | - | 2 | 263 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC728485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294306.1 | NP_001281235.1 | 2 | 297 | ||
GenBank mRNA | AK092138.1 | - | 2 | 263 |