siRNA Details
Gene Symbol: DNLZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080849.2 | NP_001074318.1 | 2, 3 | 467 | ||
NR_073565.1 | 2, 3 | 471 | ||||
GenBank mRNA | BC146915.1 | - | 2, 3 | 467 | 9056843 | |
BC146924.1 | - | 2, 3 | 467 | 9056852 |
