siRNA Details
Gene Symbol: CDK11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291345.1 | NP_001278274.1 | 20 | 2824 | ||
NM_001787.2 | NP_001778.2 | 20 | 2893 | |||
NM_033486.2 | NP_277021.2 | 20 | 2854 | |||
NM_033487.2 | NP_277022.1 | 18 | 2594 | |||
NM_033489.2 | NP_277024.2 | 21 | 2901 | |||
NM_033490.2 | NP_277025.1 | 19 | 2717 | |||
XM_006711065.3 | XP_006711128.1 | 20 | 3605 | |||
XM_011542490.2 | XP_011540792.1 | 20 | 3662 | |||
XM_011542491.2 | XP_011540793.1 | 20 | 3118 | |||
XM_011542492.2 | XP_011540794.1 | 20 | 2969 | |||
XM_011542493.2 | XP_011540795.1 | 20 | 3632 | |||
XM_011542494.2 | XP_011540796.1 | 21 | 3709 | |||
XM_011542495.2 | XP_011540797.1 | 21 | 3679 | |||
XM_011542496.2 | XP_011540798.1 | 15 | 2149 | |||
XM_017002925.1 | XP_016858414.1 | 20 | 3623 | |||
XM_017002926.1 | XP_016858415.1 | 20 | 3593 | |||
XM_017002927.1 | XP_016858416.1 | 20 | 3585 | |||
XM_017002928.1 | XP_016858417.1 | 21 | 3682 | |||
GenBank mRNA | DB516340.1 | - | 1 | 248 |
Gene Symbol: CDK11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313896.1 | NP_001300825.1 | 20 | 2967 | ||
NM_001313982.1 | NP_001300911.1 | 20 | 2955 | |||
NM_024011.3 | NP_076916.2 | 20 | 2958 | |||
NM_033529.3 | NP_277071.2 | 20 | 2928 | |||
NR_132739.1 | 20 | 2842 | ||||
NR_132740.1 | 20 | 2881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
