siRNA Details
Gene Symbol: DMRTC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080851.1 | NP_001074320.1 | 6 | 831 | ||
XM_006724684.2 | XP_006724747.1 | 7 | 910 | |||
XM_006724685.3 | XP_006724748.1 | 7 | 935 | |||
XM_017029776.1 | XP_016885265.1 | 7 | 941 |
Gene Symbol: DMRTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033053.2 | NP_149042.2 | 6 | 831 | ||
XM_005262287.3 | XP_005262344.1 | 7 | 910 | |||
XM_005262288.3 | XP_005262345.1 | 7 | 935 | |||
XM_017029725.1 | XP_016885214.1 | 7 | 941 | |||
GenBank mRNA | AV753283.1 | - | 376 | |||
BC029799.1 | - | 6 | 1255 | |||
BC047596.1 | - | 5 | 779 | 4830694 |
