siRNA Details
Gene Symbol: LOC105369246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011523514.2 | XP_011521816.1 | 1 | 1090 |
Gene Symbol: LOC400553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126852.1 | - | 2 | 1805 |
