siRNA Details
Gene Symbol: CT45A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017435.1 | NP_001017435.1 | 3 | 495 | ||
XM_006724758.3 | XP_006724821.1 | 3 | 412 | |||
XM_011531340.2 | XP_011529642.1 | 3 | 371 | |||
GenBank mRNA | AK098689.1 | - | 3 | 340 | ||
AY743711.1 | - | 3 | 495 |
Gene Symbol: CT45A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017417.2 | NP_001017417.1 | 3 | 494 | ||
XM_005278141.3 | XP_005278198.1 | 3 | 343 | |||
XM_011531352.1 | XP_011529654.1 | 2 | 264 | |||
GenBank mRNA | AY743709.1 | - | 3 | 495 | ||
BC140913.1 | - | 3 | 394 | 9056725 |
Gene Symbol: CT45A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152582.6 | NP_689795.4 | 3 | 495 | ||
XM_006724783.2 | XP_006724846.1 | 3 | 422 | |||
XM_011531389.2 | XP_011529691.1 | 3 | 390 | |||
GenBank mRNA | AY743710.1 | - | 3 | 495 |
Gene Symbol: CT45A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028711.2 | - | 3 | 495 | 4828274 | |
BC140735.1 | - | 3 | 402 | 9021603 | ||
BC140926.1 | - | 3 | 402 | 9056738 | ||
BC144437.1 | - | 3 | 402 | 9052959 |
Gene Symbol: CT45A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY743714.1 | - | 3 | 495 |
Gene Symbol: CT45A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU540102.1 | - | 3 | 314 |
Gene Symbol: CT45A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291535.1 | NP_001278464.1 | 3 | 495 | ||
XM_006724799.2 | XP_006724862.1 | 3 | 422 | |||
XM_011531241.2 | XP_011529543.1 | 3 | 390 | |||
GenBank mRNA | BP371111.1 | - | 3 | 340 |
Gene Symbol: CT45A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291540.2 | NP_001278469.1 | 3 | 495 | ||
NM_001321271.1 | NP_001308200.1 | 3 | 340 | |||
XM_006724796.2 | XP_006724859.1 | 3 | 422 |
