siRNA Details
Gene Symbol: LINC00910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027412.1 | 1 | 571 | |||
NR_027413.2 | 1 | 571 | ||||
GenBank mRNA | BC035366.1 | - | 1 | 218 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027412.1 | 1 | 571 | |||
NR_027413.2 | 1 | 571 | ||||
GenBank mRNA | BC035366.1 | - | 1 | 218 |