siRNA Details
Gene Symbol: MEIKIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303622.1 | NP_001290551.1 | 11 | 1104 | ||
GenBank mRNA | AB987829.1 | - | 11 | 911 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: BC107368, BC107369, AB987828, AK015073.
Gene Ontology Categories:
