siRNA Details
Gene Symbol: LOC729296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109916.1 | 2, 3 | 437 | |||
NR_109917.1 | 2, 3 | 437 | ||||
NR_109918.1 | 2, 3 | 437 | ||||
GenBank mRNA | DB026168.1 | - | 2, 3 | 415 | ||
DB051395.1 | - | 2, 3 | 437 | |||
HY046567.1 | - | 2, 3 | 251 |
