siRNA Details
Gene Symbol: CCDC61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267723.1 | NP_001254652.1 | 12 | 1411 | ||
XM_005259191.3 | XP_005259248.1 | 12 | 3516 | |||
XM_005259192.4 | XP_005259249.1 | 12 | 2157 | |||
XM_011527257.2 | XP_011525559.1 | 12 | 1440 | |||
GenBank mRNA | CA775840.1 | - | 4 | 349 | ||
CB852675.1 | - | 5 | 398 | |||
GQ375050.1 | - | 8 | 844 |
