siRNA Details
Gene Symbol: LOC102724467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120309.1 | 2 | 401 | |||
GenBank mRNA | DB094384.1 | - | 2 | 403 | ||
DB100335.1 | - | 2 | 448 | |||
HY133504.1 | - | 1 | 114 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102724467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120309.1 | 2 | 401 | |||
GenBank mRNA | DB094384.1 | - | 2 | 403 | ||
DB100335.1 | - | 2 | 448 | |||
HY133504.1 | - | 1 | 114 |