siRNA Details
Gene Symbol: XCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024644.1 | NP_001019815.1 | 2 | 952 | ||
NM_005283.2 | NP_005274.1 | 3 | 1074 | |||
XM_017006196.1 | XP_016861685.1 | 4 | 2092 | |||
XM_017006197.1 | XP_016861686.1 | 6 | 2344 | |||
XM_017006198.1 | XP_016861687.1 | 5 | 2157 | |||
XM_017006199.1 | XP_016861688.1 | 4 | 1154 | |||
XM_017006200.1 | XP_016861689.1 | 3 | 2012 | |||
XM_017006201.1 | XP_016861690.1 | 2 | 1861 | |||
GenBank mRNA | AK314277.1 | - | 2 | 1205 | ||
BC069075.1 | - | 1 | 850 | 7216889 | ||
BC069589.1 | - | 1 | 850 | 7262280 | ||
BC095533.1 | - | 1 | 901 | 6971949 |
