siRNA Details
Gene Symbol: ZNF600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321866.1 | NP_001308795.1 | 3 | 604 | ||
XM_017026391.1 | XP_016881880.1 | 3 | 507 | |||
GenBank mRNA | DB158392.1 | - | 2 | 454 |
