siRNA Details
Gene Symbol: LOC102724344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135182.1 | 3 | 228 | |||
NR_135183.1 | 2 | 112 | ||||
GenBank mRNA | DB046859.1 | - | 2 | 112 | ||
DB057778.1 | - | 3 | 227 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102724344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135182.1 | 3 | 228 | |||
NR_135183.1 | 2 | 112 | ||||
GenBank mRNA | DB046859.1 | - | 2 | 112 | ||
DB057778.1 | - | 3 | 227 |