siRNA Details
Gene Symbol: ZNF833P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126753.1 | - | 6 | 630 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF833P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126753.1 | - | 6 | 630 |