siRNA Details
Gene Symbol: HLA-DRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF097680.1 | - | 1 | 79 | ||
AK292140.1 | - | 2 | 242 | |||
M15992.1 | - | 1 | 92 | |||
M16955.1 | - | 1 | 92 | |||
M16956.1 | - | 1 | 92 | |||
M20503.1 | - | 2 | 242 | |||
M30181.1 | - | 2 | 179 | |||
M32578.1 | - | 2 | 240 | |||
M57648.1 | - | 2 | 237 | |||
U66826.1 | - | 2 | 219 |
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ140279.1 | - | 2 | 239 | ||
M14661.1 | - | 1 | 41 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
