siRNA Details
Gene Symbol: UQCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085372.2 | NP_001078841.1 | 2 | 598 | ||
GenBank mRNA | AY358935.1 | - | 2 | 598 | ||
BC090057.1 | - | 2 | 566 | 30347387 | ||
BG385605.1 | - | 2 | 569 |
