siRNA Details
Gene Symbol: Abcg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF323659.1 | - | 5, 6 | 703 | ||
BC119471.2 | - | 5, 6 | 655 | 40048166 | ||
U34920.1 | - | 5, 6 | 665 | |||
Z48745.1 | - | 5, 6 | 694 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
